Learn More
Applied Biosystems™ VeritiLink™ Remote Management Software
Catalog No. 4393565
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 software
Catalog No. Quantity
43-935-65 1 software
1 options
Catalog No. 43-935-65 Supplier Applied Biosystems™ Supplier No. 4393565
Add to Cart
Add to Cart
Request a Quote
Easy-to-install software provides secure, remote management for Veriti Thermal Cyclers from anywhere on network
VeritiLink Remote Management Software (VRMS) browser-enabled interface means you can “Bookmark Your Veritis” for fast, convenient access
  • Create and edit methods on your computer for increased security and peace of mind, and download/upload to/from your Veriti systems as needed
  • Perform and monitor runs for up to 50+ networked Veriti Thermal Cyclers
  • Manage all Veriti™ systems in secure client-server environment with user password protection
  • Receive real-time email system updates on desktop, laptop or PDA
  • Easy setup: install software on server for use on any networked computers
  • Veriti Systems Anywhere, Everywhere: VeritiLink Remote Monitoring Software VRMS allows you to access Veriti Systems anywhere, everywhere
  • Software package allows to manage and monitor thermal cycling runs wherever you can access an internet browser
  • With this program, you can start or stop runs, create and edit methods and send them to Veriti Thermal Cycler systems as needed
  • Designed to manage from one to 50+ Veriti Thermal Cyclers, user interface shows which systems are available for use and status of systems in use to facilitate instrument sharing for busy labs
  • With system update email notifications to your desktop, laptop, or PDA and user password protection to keep your methods secure
  • Software provides with peace of mind that systems are running smoothly and securely
  • Easy to Install: Software is easy to install, requiring only server-side installation to provide access for all networked computers
PCR, PCR & Real-Time PCR
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Veriti Thermal Cycler
Product Line VeritiLink
Software Type Remote Management Software
Product Type Thermal Cycler Accessories
For Use With (Application) PCR
Quantity 1 software
For Research Use Only. Not for use in diagnostic procedures.