Learn More
AdipoGen anti-APRIL (human), mAb (blocking) (Mahya-1) (preservative free) (500 µg)
SDP
Supplier:  AdipoGen AG20B0078PFC500
Encompass_Preferred
Monoclonal Antibody. Recognizes human APRIL. Does not recognize mouse APRIL or mouse and human BAFF. Applications: ELISA, FUNC (Blocking), IP. Isotype: Mouse IgG1 kappa. Clone: Mahya-1. Liquid. In PBS. The B cell-stimulating molecules, BAFF (B cell activating factor) and APRIL (a proliferation-inducing ligand), are critical factors in the maintenance of the B cell pool and humoral immunity. APRIL maintains B cell homeostasis by acting at a later stage, modulating the function and survival of antigen-experienced B cells. APRIL (a proliferation-inducing ligand) is implicated in several human autoimmune diseases with autoreactive B cell involvement, including systemic lupus erythematosus (SLE), Sjögren’s syndrome (SS), IgA nephropathy (IgAN) and rheumatoid arthritis (RA). APRIL might also function in enhancing proliferation of some tumor cells, especially B-cell malignancies.
Catalog No. 50-205-2992
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.