Learn More
DuPont™ Tychem™ 6000 FR Sleeved Apron
Catalog No. 19122048 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Color:
Gray
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-122-048 Gray Small
19-122-049 Gray Medium
19-122-051 Gray X-Large
19-122-052 Gray -
19-122-053 Gray 3X-Large
19-122-054 Gray 4X-Large
19-122-055 Orange Small
19-122-056 Orange Medium
19-122-057 Orange Large
19-122-058 Orange -
19-122-059 Orange 2X-Large
19-122-060 Orange 3X-Large
19-122-061 Orange 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Catalog No. 19-122-048 Supplier DuPont™ Supplier No. TP275TGYSM000200
This item is not returnable. View return policy
A single layer garment that provides triple hazard protection from liquid-chemical splash, flash fire and electric arc
  • Tychem™ Thermopro for triple hazard protection
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Provides frontal protection
  • For flash fire and electric arc hazards, partial body protective Tychem™ ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
  • Sewn with Nomex™ thread
  • Two buckle closure system
  • Open back for wearer comfort.
  • Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
  • Gray color for discretionary needs
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Material Tychem
Color Gray
Closure Type Waist Ties