Learn More
Tocris Bioscience™ Aprotinin
GREENER_CHOICE
Catalog No. 413910
Change view
Click to view available options
10mg
Catalog No.
41-391-0 10mg
1 options
This item is not returnable. View return policy
Catalog No. 41-391-0 Supplier Tocris Bioscience™ Supplier No. 4139/10
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Competitive serine protease inhibitor
Aprotinin is a competitive serine protease inhibitor. Reversibly binds to and blocks the enzymatic active site. Inhibits a range of serine proteases including trypsin, chymotrypsin, kallikrein and plasmin. Inhibits cytopathogenic effect of SARS-CoV-2 and double-stranded RNA formation in SARS-CoV-2-infected cells.
This product is from Bovine Lung.
Specifications
Molecular Weight (g/mol) 6511.48
Product Type Aprotinin
Quantity 10 mg