Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aprotinin in Solution from Bovine Lung, MP Biomedicals

Catalog No. MP219077910 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219077910 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219077910 Supplier MP Biomedicals, Inc Supplier No. 0219077910
Add to Cart
Add to Cart

  • Another commonly used unit of activity is the KIU (Kallikrein Inhibitor Unit)
  • 1 TIU ∼ 1,300 KIU

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 6511.509
Quantity 10mL
Product Type Aprotinin in solution from Bovine Lung
Content And Storage +4°C
Form Solution
Color Colorless
Purity or Quality Grade Analytical
Solubility Soluble in water (10mg/mL - completely soluble) and in aqueous buffers of low ionic strengths.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.