Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA11062
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1-1062 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1-1062 Supplier Invitrogen™ Supplier No. PA11062
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-1062 detects APS from mouse samples. PA1-1062 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~ 84 kDa protein corresponding to APS from recombinant mouse protein and 3T3L1 extracts. The PA1-1062 immunogen is a synthetic peptide corresponding to residues E(605)A T L G R A R A V E N Q Y S(619) from mouse APS. This peptide (Cat. # PEP-246) can be used in neutralization and control experiments.

APS (adapter protein with Pleckstrin homology and Src homology 2 domains) a member of the Lnk family, is an adaptor protein that is involved in B-cell signaling, insulin signaling, cytoskeletal reorganization. It is tyrosine phosphorylated by JAK2, KIT and other kinases during B-cell receptor stimulation of many different cytokines, chemokines and leukokines. APS has been shown to inhibit the JAK-STAT pathway in collaboration with c-Cbl. It is located in the cytoplasm in pre-PDGF (platelet derived growth factor) stimulated cells and localizes to the plasma membrane and peripheral regions upon PDGF stimulation. This protein is expressed in B-cells and mast cells when detected in haematopoietic cell lines yet is also expressed in many different tissues including spleen, prostrate, testis, uterus, small intestine and skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APS
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene SH2B2
Gene Accession No. Q9JID9
Gene Alias adapter protein with pleckstrin homology and Src homology 2 domains; adaptor protein with pleckstrin homology and src homology 2 domains; Alpha-1-antitrypsin; Aps; SH2 and PH domain-containing adapter protein APS; SH2B adapter protein 2; SH2B adaptor protein 2; Sh2b2
Gene Symbols SH2B2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues E(605) A T L G R A R A V E N Q Y S(619) from mouse APS.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23921
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.