Learn More
3M™ Petrifilm™ Aqua Heterotrophic Count Plates
SCI_ED
Catalog No. S30737 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Membrane filtration systems
Catalog No. For Use With (Equipment)
S30737 Membrane filtration systems
1 options
Catalog No. S30737 Supplier 3M™ Petrifilm™ Supplier No. 6450
Add to Cart
Add to Cart
Sample-ready count plates for fast, reliable detection of heterotrophic bacteria in bottled water and beverages.
  • Purpose: Detects heterotrophic bacteria in bottled water and beverages
  • Ready-to-Use: Eliminates time-consuming media and agar preparation steps
  • Results: Available within 48 to 72 hours
  • Compatibility: Designed for use with membrane filtration systems
  • Applications: Bacteria testing in beverages and bottled water
  • Product Type: Count plates
Specifications
For Use With (Application) Bacteria testing in beverages and bottled water
For Use With (Equipment) Membrane filtration systems
Type Count plates