Learn More
Electron Microscopy Sciences Aqua-Hold Pap Pen, holds 13ml of long-lasting material
SDP
Supplier:  Electron Microscopy Sciences 71311
Encompass_Preferred
The Aqua-Hold Pap Pen draws a liquid repellent barrier around a section of tissue. Used like a paint pen, you just draw a well on a slide. The barrier creates the proper surface tension to hold aqueous solutions such as costly antibody and other valuable specimens. After application, the barrier material is insoluble in alcohol and acetone but can be removed after staining with xylene. Each pen holds 13ml of long-lasting material, making hundreds of wells. Red tint distinguishes the well drawing on the slide to easily view.
Catalog No. 50-365-050
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.