Learn More
ATCC ATCC Aquabacterium commune Kalmbach et al. (BAA-209) Type strain, freeze-dried culture
Supplier:  ATCC BAA209
Encompass
Aquabacterium commune Kalmbach et al. (ATCC BAA-209) is the type strain of Aquabacterium commune, isolated from biofilms on glass and polyethylene surfaces in household drinking water supply lines. Supplied as a freeze-dried research culture for microbiology and environmental biofilm studies, the strain is aerobic and typically cultured on low-nutrient R2A medium at room temperature.
  • Type strain confirmed and characterized.
  • Isolated from drinking water supply biofilms.
  • Grow on modified R2A medium (low nutrient).
  • Incubate aerobically at 20-22 °C.
  • Supplied freeze-dried; store at 2-8 °C.
  • Biosafety level 1; standard laboratory handling precautions.
Catalog No. NC3678882