Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Aquaponics Kit
SDPSCI_ED

Catalog No. s43403 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
50 tests per factor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S43403 50 tests per factor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43403 Supplier LaMotte™ Supplier No. 3637
Add to Cart
Add to Cart

Includes

  • Detailed, diagrammed instruction manual
  • Quick-reference lid instruction

LaMotte™ Aquaponics test kit monitors basic water quality for both the aquaculture and hydroponics segments of your system

As the farm-to-table trend continues to spread, aquaponics is growing rapidly as a hobby and as an industry. The test kit is designed for small to midsize aquaculture and hydroponics systems.

The kit is complete with reagents, labware and accessories, mounted in a foam-lined carrying case, to perform approximately 50 repetitions per test factor. Test chemistries and comparators are compatible with fresh, brackish or salt water systems.

OCTA-SLIDE 2 COMPARATOR

Method:

  • Ammonia: sallicylate method
  • Nitrite: Diazotization/coupling
  • Nitrate: Zinc reduction
  • pH: Wide range indicator
  • Iron, Ferrous and Ferric: Bipyridyl indicator

Range/Sensitivity:

  • Ammonia: 0.0, 0.05, 0.1, 0.25, 0.5, 1.0, 1.5, 2.0 ppm NH3-N
  • Nitrite: 0.05, 0.1, 0.2, 0.3, 0.4, 0.5, 0.6, 0.8 ppm N2-N
  • Nitrate: 0.25, 0.5, 1.0, 2.0, 4.0, 6.0, 8.0, 10.0 ppm NO3-N
  • pH: 5.0, 6.0, 7.0, 7.5, 8.0, 9.0, 10.0 pH
  • Iron, Ferrous and Ferric: 0.5, 1.0, 2.0, 3.0, 4.0, 6.0, 8.0, 10.0 ppm Fe

DIRECT READING TITRATOR

Method:

  • Alkalinity: Acid/base titration
  • Dissolved Oxygen: Modified Winler titration

Range/Sensitivity:

  • Alklinity: 0 to 200ppm as CaCO3, in 4.0 ppm increments
  • Dissolved oxygen: 0 to 10 ppm in 0.2 ppm

Specifications

For Use With (Application) Aquaculture and hydroponics
Includes Instruction manual and quick-reference lid instruction
Quantity 50 tests per factor
Product Type Water Quality Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.