Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aquaporin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553234 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553234 Supplier Invitrogen™ Supplier No. PA553234
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CPDVEFKRRF KEAFSKAAQQ TKGSYMEVED NRSQVETDDL ILKPGVVHVI DVDRGEEKKG KDQSGEVLSS V Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

In skeletal muscle, AQP4 (aquaporin 4 also known as mercurial insensitive water channel), localizes to the sarcolemma of fast-twitch muscle fibers. Aquaporins (AQPs) are a large family of integral membrane water transport channel proteins that facilitate the transport of water through the cell membrane. This function is conserved in animals, plants and bacteria. Many isoforms of aquaporin have been identified in mammals, designated AQP0 through AQP10. Aquaporins are widely distributed and it is not uncommon for more than one type of AQP to be present in the same cell. Although most aquaporins are only permeable to water, AQP3, AQP7, AQP9 and one of the two AQP10 transcripts are also permeable to urea and glycerol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 4
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AQP4
Gene Accession No. P55087, P55088
Gene Alias Aqp4; AQP-4; aquaporin 4; aquaporin type4; Aquaporin4; aquaporin-4; HMIWC2; Mercurial-insensitive water channel; MGC22454; MIWC; MIWC2; WCH4
Gene Symbols AQP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Aquaporin 4. Recombinant protein control fragment (Product #RP-91109).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11829, 361
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.