Learn More
Invitrogen™ Aquaporin 5 Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Lung Tissue, Rat Testis Tissue. IHC: Rat Lung Tissue.
Specifications
Specifications
| Antigen | Aquaporin 5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal |
| Gene | AQP5 |
| Gene Accession No. | P47864, Q9WTY4 |
| Gene Alias | AQP 5; Aqp5; AQP-5; aquaporin 5; Aquaporin5; aquaporin-5; PPKB |
| Gene Symbols | AQP5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.