Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aquaporin 5 Recombinant Rabbit Monoclonal Antibody (10V2Y1)

Catalog No. MA569073
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA569073 20 μL
FEMA551429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA569073 Supplier Invitrogen™ Supplier No. MA569073
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen Sequence: IGLSVTLGHL VGIYFTGCSM NPARSFGPAV VMNRFSPAHW VFWVGPIVGA VLAAILYFYL LFPNSLSLSE RVAIIKGTYE PDEDWEEQRE ERKKTMELTT R.

Many isoforms of aquaporin have been identified in mammals, designated AQP0 through AQP10. Aquaporins are widely distributed and it is not uncommon for more than one type of AQP to be present in the same cell. Although most aquaporins are only permeable to water, AQP3, AQP7, AQP9 and one of the two AQP10 transcripts are also permeable to urea and glycerol. AQP2 is the only water channel that is activated by vasopressin to enhance water reabsorption in the kidney collecting duct. Aquaporins are involved in renal water absorption, generation of pulmonary secretions, lacrimation, and the secretion and reabsorption of cerebrospinal fluid and aqueous humor. In the lung, AQP5 is responsible for the majority of water transport across the apical membrane of type I alveolar epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 5
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 10V2Y1
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene AQP5
Gene Accession No. P47864, P55064, Q9WTY4
Gene Alias AQP 5; Aqp5; AQP-5; aquaporin 5; Aquaporin5; aquaporin-5; PPKB
Gene Symbols AQP5
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11830, 25241, 362
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.