Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AQUApure Tri-Ub Chains (K29-linked), Carrier-Free Protein

Catalog No. UC85025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
UC85025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. UC85025 Supplier R&D Systems™ Supplier No. UC85025
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The AQUApure Tri-Ub Chains (K29-linked) Protein is derived from E. coli. The AQUApure Tri-Ub Chains (K29-linked) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P0CG47
For Use With (Application) Bioactivity
Gene ID (Entrez) 7314
Molecular Weight (g/mol) Theoretical M.W.: 26kDa
Quantity 25 μg
Source E. coli-derived human Tri-Ubiquitin protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Common Name Tri-Ubiquitin
Gene Symbol UBB
Biological Activity Ubiquitin chains vary in length, linkage, and function. K29-linked Tri-Ubiquitin Chains (Ub3) are ideal for investigating Ubiquitin-binding proteins and as substrates for Ubiquitin-specific isopeptidases. Reaction conditions will need to be optimized for each specific application. IMPORTANT: Heating this product in SDS-PAGE buffer or terminating reactions containing this product with heated SDS-PAGE buffer could lead to unexpected, high apparent molecular weight banding or smearing on gels that is not representative of product purity. For optimal results, we recommend incubation in SDS-PAGE buffer + DTT at <40°C for 20 minutes prior to gel electrophoresis.
Conjugate Unconjugated
Species Human
Expression System E. coli-derived
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. Ub-AQUA analysis: K29: 99.53% K6 : 0.24% K48: 0.17% All other linkages ≤ 0.04%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.