Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AR-M 1000390 hydrochloride, Tocris Bioscience™
SDP

Catalog No. 433550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-355-0 50 mg
43-351-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-355-0 Supplier R&D Systems Supplier No. 4335/50
This item is not returnable. View return policy

Low-internalizing δ agonist

Non-peptidic, low-internalizing δ-selective opioid receptor agonist; derivative of SNC 80 (Cat. No. 0764). Does not trigger acute desensitization of the analgesic response; reduces CFA-induced hyperalgesia. Brain penetrant following systemic administration.

Chemical Identifiers

CAS 209808-47-9
Molecular Formula C23H29ClN2O
Molecular Weight (g/mol) 384.948
InChI Key OTXTZCLQEGSAMW-UHFFFAOYSA-N
Synonym ar-m 1000390 hydrochloride, arm390 hydrochloride, arm-390hcl, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl-benzamide hydrochloride, n,n-diethyl-4-phenyl piperidin-4-ylidene methyl benzamide hydrochloride, arm1000390 hydrochloride, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl benzamide hydrochloride
PubChem CID 76848958
IUPAC Name N,N-diethyl-4-[phenyl(piperidin-4-ylidene)methyl]benzamide;hydrochloride
SMILES CCN(CC)C(=O)C1=CC=C(C=C1)C(=C2CCNCC2)C3=CC=CC=C3.Cl

Specifications

Quantity 50 mg
Formula Weight 384.94
Percent Purity >97%
Chemical Name or Material AR-M 1000390 hydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.