Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AR420626 50mg | 1798310-55-0 | 417.29 g/mol | C21H18Cl2N2O3 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY11652250MG

Encompass_Preferred

AR420626 is a selective agonist of free fatty acid receptor 3 (FFAR3/GPR41) used as a research compound. It shows activity at FFAR3 (IC50 = 117 nM) and has reported anti-inflammatory, anticancer, and antidiabetic effects in preclinical studies. Supplied as a solid for laboratory use.

  • Selective FFAR3 agonist (IC50 = 117 nM).
  • High purity (98.29%).
  • Molecular weight 417.29 g/mol.
  • Chemical formula C21H18Cl2N2O3.
  • Provided as a solid research reagent, available as a 50 mg quantity.

Catalog No. 50-002-27262


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.