Learn More
Medchemexpress LLC AR420626 5mg | 1798310-55-0 | 417.29 g/mol | C21H18Cl2N2O3 | 5 MG
Supplier: Medchemexpress LLC HY1165225MG
AR420626 is a research small molecule that functions as a selective agonist of free fatty acid receptor 3 (FFAR3/GPR41). It is used in preclinical studies to probe FFAR3-mediated signaling and biological effects, including anti-inflammatory, antidiabetic, and anticancer responses. Supplied as an off-white to light yellow solid in research-scale quantities.
- Selective FFAR3 agonist with reported nanomolar activity.
- Shown to modulate inflammatory and metabolic pathways in preclinical models.
- Available in small pack sizes suitable for in vitro and in vivo studies.
- Off-white to light yellow solid appearance for easy handling and verification.
- Recommended storage: keep powder at -20°C for long-term stability.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.