Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ara H1 Recombinant Mouse Monoclonal Antibody (10C4)

Catalog No. MA566231
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA566231 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA566231 Supplier Invitrogen™ Supplier No. MA566231
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Recombinant Monoclonal Antibody

Ara H1 Recombinant Monoclonal Antibody for ELISA

Ara h 1 is a major allergen found in peanuts (Arachis hypogaea) and is classified as a seed storage protein. It belongs to the vicilin family of 7S globulins and is known for its strong allergenic potential, contributing significantly to peanut allergy, which is among the most common food allergies. Structurally, Ara h 1 is a glycoprotein with a molecular weight of approximately 63.5 kDa, consisting of a trimeric formation that enhances its stability and resistance to digestion. This resistance contributes to its allergenicity, as it can persist in the gastrointestinal tract and elicit immune responses. Immunologically, Ara h 1 contains multiple epitopes recognized by IgE antibodies in susceptible individuals, triggering allergic reactions. Due to its stability and prevalence, Ara h 1 is a key target in diagnostic tests and immunotherapies aimed at desensitizing allergic patients to peanuts. Ongoing research includes characterizing its structure, identifying IgE-binding sites, and understanding its role in mediating allergic responses to develop effective treatments and potentially hypoallergenic peanut varieties.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ara H1
Applications ELISA
Classification Recombinant Monoclonal
Clone 10C4
Concentration 5.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Host Species Mouse
Immunogen Peanut allergen Ara H1.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.