Learn More
Cayman Chemical Arachidonic Acid, 506-32-1, 500 mg
Supplier: Cayman Chemical 90010500
Molecular Formula C20H32O2, Purity ≥98%, Formula Weight 304.5, Synonyms: 5Z,8Z,11Z,14Z-eicosatetraenoic acid, AA; Keystone essential fatty acid that is converted by COX, LO, and epoxygenase enzymes into more than 150 potent metabolites in species ranging from fungi to plants to mammals.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.