Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arachidonic Acid, MP Biomedicals

Catalog No. MP219462550 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219462550 50 mg
MP219462590 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219462550 Supplier MP Biomedicals, Inc Supplier No. 0219462550

  • If ethanol is undesirable, arachidonic acid may be dissolved in acetonitrile, DMF, or DMSO
  • Simply evaporate the ethanol under a gentle stream of nitrogen (be certain not to evaporate the material to dryness) and redissolve the arachidonic acid in the solvent of choice
  • Just prior to use, make dilutions of the stock solution into aqueous buffer or isotonic saline to bring the arachidonic acid to the desired concentration
  • Ensure that the residual amount of organic solvent is insignificant, since organic solvents may have physiologic effects at low concentrations
  • A control using the solvent in the absence of the prostaglandin will address this potential variable
  • We do not recommend storing the aqueous solution for more than one day
  • It is difficult to obtain aqueous solutions of arachidonic acid directly
  • However, an organic solvent free solution of arachidonic acid can be prepared using concentrated basic buffers (pH > 8.0 and ionic strength not less than 0.1 M). Add 400 µL of cold buffer (0°C) per mg of arachidonic acid and agitate vigorously and/or ultrasonicate

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-32-1
Molecular Formula C20H32O2
Molecular Weight (g/mol) 304.474
MDL Number MFCD00004417
InChI Key YZXBAPSDXZZRGB-DOFZRALJSA-N
Synonym arachidonic acid, arachidonate, immunocytophyte, all-z-5,8,11,14-eicosatetraenoic acid, 5z,8z,11z,14z-icosa-5,8,11,14-tetraenoic acid, cis-5,8,11,14-eicosatetraenoic acid, 5,8,11,14-eicosatetraenoic acid, all-z, all-cis-5,8,11,14-eicosatetraenoic acid, unii-27yg812j1i, 5z,8z,11z,14z-eicosatetraenoic acid
PubChem CID 444899
ChEBI CHEBI:15843
IUPAC Name (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoic acid
SMILES CCCCCC=CCC=CCC=CCC=CCCCC(=O)O

Specifications

Melting Point -49°C (literature)
Color Colorless
Boiling Point 170°C (0.2 mmHg (literature))
Linear Formula C20H32O2
Quantity 50 mg
Solubility Information Soluble in chloroform(50mg/mL) and absolute ethanol (10mg/mL)(Lit.)
Formula Weight 304.5
Percent Purity ≥98%
Grade Cell Culture Reagent
Physical Form Liquid
Chemical Name or Material ARACHIDONIC ACID
Show More Show Less
Recommended Storage -20°C. Store Desiccated. Protect from light. Store under nitrogen.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.