Learn More
Cayman Chemical Arachidonic Acid (peroxide free), 506-32-1, 25 mg
SDP
Supplier:  Cayman Chemical 90010.125
Encompass_Preferred
Free arachidonic acid is a transient, critical substrate for the biosynthesis of eicosanoid second messengers. Arachidonic acid oxidizes spontaneously on exposure to room air, yielding a complex mixture of fatty acid peroxides. Peroxide-free arachidonic acid is obtained after removal of the peroxides by silicic acid chromatography, followed by immediate addition of the antioxidant BHT (2,6-Di-tert-butyl-4-hydroxytoluene). Arachidonic acid is also available without BHT (Catalog No. 90010).
Catalog No. NC1041945
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.