Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Aramid Fiber Cut-Resistant Glove Liners

Catalog No. 19048678 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048-678 Small
19-048-679 Medium
19-170-408C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-048-678 Supplier Wells Lamont™ Supplier No. M79S
This item is not returnable. View return policy

Liners provide ANSI Cut Level A2 protection and can be worn alone, or used as a liner for additional protection.

  • Aramid Fibers provide ANSI Cut Level A2 protection
  • Lightweight design provides cut resistance without compromising tactile sensitivity
  • Wear alone, or under another pair of gloves for additional protection
  • Economical
  • Low linting
  • Made in US

Specifications

Coating Material Uncoated
Color Yellow
Cut Resistance Rating ANSI A2
Hand Ambidextrous
Laboratory/Cleanroom Use Yes
Latex Free Yes
Material Aramid Fiber
Size Small
For Use With (Application) Critical Environments, Cleanrooms, and Electronics Assembly
Glove Cuff Style Knit
Hem Color White
Series M79
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.