Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Aramid Fiber 1800 Medium Weight Gloves

Catalog No. 17200372 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Glove Cuff Color:
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
17-200-372 Medium Yellow
17-200-371 Small Yellow
17-200-373 Large Yellow
17-200-374 X-Large Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-200-372 Supplier Wells Lamont™ Supplier No. 1800M
This item is not returnable. View return policy

Medium weight gloves are constructed of 100% aramid fiber and offer ANSI Level A2 protection.

  • 100% aramid fiber
  • Ambidextrous
  • Made in US

Specifications

ANSI Abrasion Rating ANSI 1
Certifications/Compliance ASTM F1790-97, ASTM D3884-05
Construction String Knit
Coating Material Uncoated
Color Yellow
Cut Resistance Rating ANSI A2
Hand Ambidextrous
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) Steel processing, metal handling
Glove Cuff Color Yellow
Model 1800M
Series Medium Weight 1800
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.