Learn More
Wells Lamont™ Aramid Fiber Light/Medium Weight PVC Dotted Glove
Shop All Wells Lamont Industry Group ProductsDescription
- 100% Kevlar™ Aramid fibers
- PVC dots on both sides for grip
- ambidextrous
- made in the US
Specifications
Specifications
| Color | Yellow |
| Cut Resistance Rating | ANSI A2 |
| Hand | Ambidextrous |
| Material | Aramid Fiber |
| Product Type | Cut-resistant Gloves |
| Size | Small |
| Glove Cuff Color | Yellow |