Learn More
Wells Lamont™ Grain Goatskin Leather Drivers Gloves with TPR Padding
Shop All Wells Lamont Industry Group ProductsDescription
- Grain goatskin leather for comfort and durability
- Aramid fiber liner provides ANSI A3 cut resistance
- TPR padding on the back of the hand protects against impacts
- Straight thumb
Specifications
Specifications
| Back Material | TPR Padding |
| Color | Natural |
| Cut Resistance Rating | ANSI A3 |
| Hand | Specific |
| Material | Grain Goatskin Leather |
| Product Type | Driver Gloves |
| Size | Large |
| Liner Material | Aramid Fiber |