Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Aramid Fiber with Latex Coated Palm Gloves

Catalog No. 19148701 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Medium
Hem Color:
Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hem Color
19-148-701 Medium Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-148-701 Supplier Wells Lamont™ Supplier No. Y9245M

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Specifications

Color Blue, Yellow
Cut Resistance Rating ANSI A2
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) Construction, recycling, lumber and mill works and landscaping
Glove Cuff Color Blue
Glove Cuff Style Knit
Hem Color Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.