Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ArC™ Amine Reactive Compensation Bead Kit (for use with LIVE/DEAD™ Fixable dead cell stain kits)

Catalog No. 501136946
Encompass_Preferred
Change view
Click to view available options
No. of Tests:
20
100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
50-113-6946 100
50-112-1496 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-113-6946 Supplier Invitrogen™ Supplier No. A10346
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ArC™ Amine Reactive Compensation Bead Kit (for use with LIVE/DEAD™ Fixable dead cell stain kits)

ArC™ Amine Reactive Compensation Bead Kit is a bead-based compensation tool specifically optimized for use with LIVE/DEAD™ Fixable dead cell stain kits.

  • Ready-to-use control
  • Eliminates the hassle of heat-treating cells as a control, saving precious sample
  • Enables accurate and consistent results

    View a selection guide for all fixable viability dyes for flow cytometry.

    Ready-to-use control
    Unlike other methods that require sample preparation to make compensation controls using cellular material, the ArC™ Amine Reactive Compensation Bead Kit is ready to use out of the dropper bottle. The kit includes two types of specially modified polystyrene microspheres that have been optimized to react with the amine-reactive dye in the LIVE/DEAD™ Fixable dead cell stain kits. The ArC™ reactive beads (Component A) bind any of the amine-reactive dyes and provide a positive signal; the ArC™ negative beads (Component B) have no reactivity and provide a negative compensation control. This ready-to-use product preserves the original sample material for experiments; precious sample is not used for the creation of compensation controls.

    Accurate and consistent results
    Unlike biological controls, which can have variable expression levels depending upon disease or development state, the Arc™ beads provide a bright, consistent signal (see figure below). Consistency in the staining pattern of compensation controls is important for obtaining the right compensation value. If the signal varies with the control material used, the compensation value can be affected, leading to erroneous results.

    How it works
    One drop of beads is added to a test tube containing LIVE/DEAD™ Fixable dead cell stain. After incubation with amine-reactive dye, the beads are washed in staining buffer, a drop of negative control beads is added (if required), and the beads are resuspended and analyzed by flow cytometry. The two components provide a distinct positive bead that is at least 50-fold brighter than the negative beads, which can be used to set compensation.

    Other compensation products
    Besides the ArC™ Amine Reactive Compensation Beads, Life Technologies also offers the AbC™ Anti-Mouse Bead Kit for capture of mouse anti-human antibodies and AbC™ Anti-Rat/Hamster Bead Kit for capture of rat anti-mouse or hamster anti-mouse antibodies.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Diameter (Metric) 6 μm
    Content And Storage Contains 1 vial ArC™ amine reactive beads and 1 vial of negative beads.
  • Store at 2°C to 8°C
  • Format Tube
    For Use With (Equipment) Flow Cytometer
    No. of Tests 100
    Quantity 1 Kit
    Detection Method Fluorescence
    Excitation Wavelength Range any lasers (UV to 633/635)
    Stain Configuration Surface
    Shipping Condition Room Temperature
    Product Line CompenFlow, Molecular Probes
    Product Type Compensation Beads
    Show More Show Less
    Are the vials of ArC Amine Reactive Compensation Bead Kit (for use with LIVE/DEAD Fixable dead cell stain kits) (Cat. No. A10346, A10648) compatible with Vaporized Hydrogen Peroxide (VHP) sterilization?

    While the polypropylene cap is compatible with Vaporized Hydrogen Peroxide (VHP) sterilization, we have not tested the compatibility of the Low-Density Polyethylene (LDPE) / Linear Low-Density Polyethylene (LLDPE) vials with VHP sterilization.

    What is the stability for the ArC Amine Reactive Compensation Bead Kit (for use with LIVE/DEAD Fixable dead cell stain kits)?

    When stored as directed (2-8 degrees C), this kit is stable for at least 1 year. Please do not freeze the kit. Please visit the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/arc_amine_reactive_comp_bead_kit_man.pdf) for further technical details.

    My ArC Amine Reactive Compensation beads are excessively clumped and cannot be resuspended by vortexing or sonication. What may have caused this?

    This can occur if the solution of beads was ever frozen. The formation of ice excludes the beads and compresses the beads together. Once frozen, the beads cannot be resuspended into solution and are no longer usable.

    For the ArC Amine Reactive Compensation Bead Kit, why must I use freshly prepared amine-reactive dye?

    This is because succinimidyl ester (NHS ester) groups are very labile to water; they dissociate readily from the dye structure if held in solvents that are contaminated with water, as is possible from condensation or absorbing water from the atmosphere.

    Besides the Live/Dead Fixable Dead Cell stain, may I conjugate other amine-reactive dyes to the beads in the ArC Amine Reactive Compensation Bead Kit?

    Yes. The succinimidyl ester (NHS ester) and isothiocyanate (e.g., FITC, TRITC) dye derivatives may be attached to the ArC beads, but you would have to optimize the amount of reactive dye per ArC beads.

    I am performing flow cytometry. My compensation matrix has values over 100. That's not possible is it?

    It is possible. But if you have a lot of values over 100, you probably need to look at the voltages that you are using for your data collection.

    I’ve been told that I need to compensate my data for flow cytometry analysis. What does that mean?

    By running single-color controls, it is possible to remove signal of one fluorophore that spills over into the collection channel for another fluorophore.

    What kind of controls do I need for flow cytometry?

    For compensation, you need to prepare a singly stained sample (or compensation beads) for each color parameter that you are using. In addition, we recommend that you use FMO (flow minus one) controls. These are controls in which you label cells or beads with every color in your panel, omitting one. Make one FMO control for each color. These controls are important for helping you properly set gates on your data.

    Do I need compensation controls for non-antibody reagents?

    Yes, you should have a compensation control for each color in your assay. For amine-reactive stains such as our LIVE/DEAD Fixable Dead Cell Stain Kits, you can label beads to make compensation controls using our ArC Amine Reactive Compensation Bead Kit (Cat. No. A10346). For other stains, such as cell cycle stains, you need to use labeled cells for your compensation control.

    Why should I worry about compensation in flow cytometry analysis?

    In a perfect world, the fluorescence emission profile for each individual fluorophore would be a very intense, narrow peak, well separated from all other emission peaks. In reality, organic dyes and fluorescent proteins have broad emission peaks, and compensation must be employed (during or after data acquisition) to correctly assign fluorescence signal to each fluorophore. Compensation is important because it removes fluorescent signal from overlapping spectra so you know that the signal you see is only the signal from the fluorophore of interest.

    How do I make compensation controls for antibodies?

    All that you need for compensation controls is an unstained sample of your cells (or negative control beads) and samples with each of your fluorophores - one per tube. You want the single-stained controls to be as bright as you expect your brightest sample to be. Antibodies can be bound to beads instead of cells using our AbC Total Antibody Compensation Bead Kit (Cat. Nos. A10513 and A10497).


    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.