Learn More
Honeywell Salisbury™ Arc Flash Kit
Catalog No. 19139208 Shop All WH Salisbury & Co. Products
Encompass_Preferred
Change view
Click to view available options
Arc Flash Rating:
8 cal/cm²
Description:
46 to 48 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back packs and safety Glasses
50 to 52 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back pack and safety Glasses
Includes: 42 to 44 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back pack and safety Glasses
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arc Flash Rating Description Size
19-139-208 8 cal/cm² 46 to 48 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back packs and safety Glasses X-Large
19-139-207 8 cal/cm² Includes: 42 to 44 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back pack and safety Glasses Large
19-139-209 8 cal/cm² 50 to 52 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back pack and safety Glasses 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-139-208 Supplier Honeywell Salisbury™ Supplier No. SKCA8XLBP
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Specifications
Arc Flash Rating 8 cal/cm²
Description 46 to 48 in. Chest navy blue coveralls, AS1000HAT, AFHOOD, Salisbury back packs and safety Glasses
Color Navy
Certifications/Compliance NFPA 70E™ 2009, CE 0120
Material Cotton
Size X-Large