Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Arc Flash Labels - Self-sticking Polyester

Catalog No. 19104671 Shop All Brady Corporation Products
Change view
Click to view available options
Display Text:
DANGER Arc Flash And Shock Hazard Appropriate PPE Required
DANGER Arc Flash And Shock Hazard Appropriate PPE Required (with picto)
WARNING Arc Flash And Shock Hazard Appropriate PPE Required
WARNING Arc Flash And Shock Hazard Appropriate PPE Required (with picto)
Quantity:
100/Roll
5 Pk.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text Quantity
19-104-671 DANGER Arc Flash And Shock Hazard Appropriate PPE Required (with picto) 5 Pk.
19-104-670 WARNING Arc Flash And Shock Hazard Appropriate PPE Required (with picto) 5 Pk.
19-104-672 WARNING Arc Flash And Shock Hazard Appropriate PPE Required 5 Pk.
19-104-673 DANGER Arc Flash And Shock Hazard Appropriate PPE Required 5 Pk.
19-104-676 WARNING Arc Flash And Shock Hazard Appropriate PPE Required (with picto) 100/Roll
19-104-678 WARNING Arc Flash And Shock Hazard Appropriate PPE Required 100/Roll
19-104-677 DANGER Arc Flash And Shock Hazard Appropriate PPE Required (with picto) 100/Roll
19-104-679 DANGER Arc Flash And Shock Hazard Appropriate PPE Required 100/Roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-104-671 Supplier Brady™ Supplier No. 101951
Add to Cart
Add to Cart

Self-sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.

  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F).
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings.
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes.
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C).

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance NEC, NFPA 70E, OSHA 1910.331-335
Description Self-adhesive Label
Adhesive Type Permanent Acrylic
Color Black, Red on White
Finish Gloss
Format Label
Material Polyester
Printable Sides Single-side Printable
Product Type Label
Sterility Non-sterile
Display Text DANGER Arc Flash And Shock Hazard Appropriate PPE Required (with picto)
Includes Pack of 5 Labels
Languages English
Quantity 5 Pk.
Shape Rectangle
Size (English) 5 x 3.5 in. (W x H)
Thickness (English) 6 mil
Thickness (Metric) 0.152 mm
Dimensions (L x W) 3.5 x 5 in.
Width (English) 5 in.
Width (Metric) 127.00 mm
Height (English) 3.5 in.
Height (Metric) 88.90 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.