Learn More
National Safety Apparel™ Arc H2O Bib Flo. Orange/Red Custom Class E
SDP
Catalog No. 170012218 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2218 3X-Large
17-001-2213 Small
17-001-2214 Medium
17-001-2215 Large
17-001-2216 X-Large
17-001-2217 2X-Large
17-001-2219 4X-Large
17-001-2220 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-2218 Supplier National Safety Apparel™ Supplier No. R40RQ193X
Add to Cart
Add to Cart
Specifications
Closure Type Buckle
Color Fluorescent Orange
Material Polyurethane-coated Cotton
Seam Type Welded
Size 3X-Large
Ankle Style Open
Arc Flash Rating 19 cal/cm² (Fluorescent Yellow); 24 cal/cm² (Fluorescent Orange)
Certifications/Compliance CAT 2, NFPA 70 E, ANSI/ISEA 107 Type R Class 3, CSA Z462, ASTM F2733, ASTM F1891, OSHA 1910.269
Disposable Reusable