Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Arc H2O™ FR Rainwear Kit - Type R Class 3
SDP

Catalog No. 170012024 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2024 5X-Large
17-001-2017 Small
17-001-2018 Medium
17-001-2019 Large
17-001-2020 2X-Large
17-001-2021 3X-Large
17-001-2022 X-Large
17-001-2023 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-2024 Supplier National Safety Apparel™ Supplier No. KITRLC35X
Add to Cart
Add to Cart

Includes Type R Class 3 FR jacket, Class E FR bib overall, and black mesh gear bag

  • 2 in. FR silver reflective trim
  • FR polyurethane (PU) moisture barrier
  • Lightweight and stretchable comfort
  • FR polyurethane is durable and flexible that will not become brittle as it ages
  • Waterproof welded seams with reinforced stress points to provide durability
  • Back ventilation system with FR mesh for increased airflow
  • Contoured collar with tuck away attached hood
  • Covered D-Ring opening access
  • Covered zipper and snap closure
  • FR hook and loop adjustable leg closures
  • Adjustable cuffs and shoulder straps
  • Heavy duty quick release suspender system
  • FR fabric will not melt, drip, or ignite

Specifications

Arc Flash Rating 8 cal/cm²
Certifications/Compliance CAT 2, NFPA 70E, ANSI/ISEA 107 Type R Class 3, CSA Z462, ASTM F2733, ASTM F1891, OSHA 1910.269
Product Type Arc Rainwear Kit
For Use With (Application) Utilities, Electrical Contractors
Gender Unisex
Color Fluorescent Yellow
Suspender Adjustable
Size 5X-Large
Ankle Style Open
Material Polyurethane-coated FR Cotton
Seam Type Welded
Closure Type Zipper, Snap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.