Learn More
National Safety Apparel™ Arc H2O Jacket FL Orange Segmented Reflective Trim Tall
Shop All National Public Seating Corporation Products
Specifications
Specifications
| Size | Medium |
| Gender | Unisex |
| Material | Polyurethane-coated FR Cotton |
| Color | Fluorescent Orange |
| Closure Type | Snap, Zipper |
| Seam Type | Welded |
| Product Type | Hi-vis Jacket |
| Length (English) | 30.5 in. |