Learn More
National Safety Apparel™ Arc H2O Jacket FL Orange Segmented Reflective Trim Tall
SDP
Catalog No. 170012128 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2128 2X-Large
17-001-2125 Small
17-001-2126 Medium
17-001-2127 Large
17-001-2129 X-Large
17-001-2130 3X-Large
17-001-2131 4X-Large
17-001-2132 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-2128 Supplier National Safety Apparel™ Supplier No. R30RQCX22XT
Add to Cart
Add to Cart
Specifications
Size 2X-Large
Gender Unisex
Material Polyurethane-coated FR Cotton
Color Fluorescent Orange
Closure Type Snap, Zipper
Certifications/Compliance Arc Rating: 19 cal/cm² (Fluorescent Yellow); 24 cal/cm² (Fluorescent Orange), CAT 2, NFPA 70 E, ANSI/ISEA 107 Type R Class 3, CSA Z462, CSA Z96 x Calss 2, ASTM F2733, ASTM F1891, OSHA 1910.269
Seam Type Welded
Product Type Hi-vis Jacket
Length (English) 30.5 in.