Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 12 Cal Long Coat/Legging Kit with Balaclava/Voltage Gloves (10)
SDP

Catalog No. 170011186 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1186 X-Large
17-001-1171 Small
17-001-1176 Medium
17-001-1181 Large
17-001-1191 2X-Large
17-001-1196 3X-Large
17-001-1201 4X-Large
17-001-1206 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1186 Supplier National Safety Apparel™ Supplier No. KIT2LC11XL10
Add to Cart
Add to Cart

Ultimate HRC 2 kit without balaclava and gloves

  • Coveralls
  • Hard Hat with ArcGuard™ Faceshield & Chin Cup
  • Safety Glasses
  • Faceshield unit bag
  • Gear bag

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 2, NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Arc Flash Rating 12 cal/cm²
Material Westex™ UltraSoft™
Size X-Large
Ankle Style Open
Closure Type FR Hook and Loop
Seam Type Stitched
Color Navy
Suspender None
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.