Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 25 cal Arc Flash Kit with Voltage Gloves (12)
SDP

Catalog No. 170011388 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1388 Large
17-001-1378 Small
17-001-1383 Medium
17-001-1393 2X-Large
17-001-1398 3X-Large
17-001-1403 4X-Large
17-001-1412 5X-Large
17-001-1413 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1388 Supplier National Safety Apparel™ Supplier No. KIT3SC25LG12
Add to Cart
Add to Cart

ArcGuard™ Arc Flash Kit with Jacket

  • Faceshield Bag
  • Faceshield with hood and hard hat
  • Glove Bag
  • Large Gear Bag
  • Leather Protectors
  • Rubber Voltage Gloves
  • Safety Glasses
  • Bib Overall
  • Short Coat

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 3, NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Product Type Arc Flash Kit
Arc Flash Rating 25 cal/cm²
Suspender Adjustable
Material Cotton, Nylon
Ankle Style Open
Closure Type FR Hook and Loop
Color Khaki
Seam Type Stitched
Size Large
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.