Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 25 cal Coat/Bib Kit, No Gloves
SDP

Catalog No. 170011418 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1418 2X-Large
17-001-1414 Small
17-001-1415 Medium
17-001-1416 Large
17-001-1417 X-Large
17-001-1419 3X-Large
17-001-1420 4X-Large
17-001-1421 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1418 Supplier National Safety Apparel™ Supplier No. KIT3SC25NG2X
Add to Cart
Add to Cart

Arc flash kit with double layer Ultrasoft™

  • Faceshield Bag
  • Faceshield with hood and hard hat
  • Large Gear Bag
  • Safety Glasses
  • UltraSoft™ Bib Overall
  • UltraSoft™ Short Coat

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 3, NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Product Type Arc Flash Kit
Arc Flash Rating 25 cal/cm²
Size 2X-Large
Suspender Adjustable
Ankle Style Open
Closure Type FR Hook and Loop
Material Cotton, Nylon
Seam Type Stitched
Color Khaki
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.