Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ 40 cal ArcGuard™ Contractor Arc Flash Kit with Short Coat and Bib Overall (10)
SDP

Catalog No. 170011694 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1694 4X-Large
17-001-1664 Small
17-001-1669 Medium
17-001-1674 Large
17-001-1679 X-Large
17-001-1684 2X-Large
17-001-1689 3X-Large
17-001-1699 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1694 missing translation for 'mfr' National Safety Apparel™ missing translation for 'supplierNo' KIT4SC40EC4X10
Add to Cart
Add to Cart

40 cal arc flash kit with short coat and bib overalls with voltage gloves

  • ArcGuard™ Economy™ Short Coat and Bib Overall
  • 14 in. Class 2 Rubber Insulating Gloves
  • 12 in. Leather Glove Protectors
  • Arc Flash Hood Unit with faceshield & hard hat
  • Safety Glasses
  • Hood bag
  • Glove bag
  • Gear bag

Specifications

Gender Unisex
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Arc Flash Rating 40 cal/cm²
Closure Type FR Hook and Loop
Seam Type Stitched
Ankle Style Open
Material Cotton, Nylon
Color Tan
Size 4X-Large
Suspender Adjustable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.