Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ Compliance 40 Cal Coverall Kit with Standard Hood and Voltage Gloves (08)
SDP

Catalog No. 170011452 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1452 4X-Large
17-001-1422 Small
17-001-1427 Medium
17-001-1432 Large
17-001-1437 X-Large
17-001-1442 2X-Large
17-001-1447 3X-Large
17-001-1457 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1452 Supplier National Safety Apparel™ Supplier No. KIT4CV404X08
Add to Cart
Add to Cart

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Material Cotton, Nylon, Polyester
Arc Flash Rating 40 cal/cm²
Size 4X-Large
Suspender Adjustable
Ankle Style Open
Closure Type FR Hook and Loop
Color Navy
Seam Type Stitched
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.