Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ Compliance 40 Cal Coverall Kit with Standard Hood and Voltage Gloves (09)
SDP

Catalog No. 170011423 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1423 Small
17-001-1428 Medium
17-001-1433 Large
17-001-1438 X-Large
17-001-1443 2X-Large
17-001-1448 3X-Large
17-001-1453 4X-Large
17-001-1458 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1423 Supplier National Safety Apparel™ Supplier No. KIT4CV40SM09
Add to Cart
Add to Cart

Specifications

Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Product Type Arc Flash Kit
Gender Unisex
Material Cotton, Nylon
Arc Flash Rating 40 cal/cm²
Closure Type FR Hook and Loop
Color Navy
Seam Type Stitched
Suspender Adjustable
Ankle Style Open
Size Small
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.