Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 40 cal Long Coat/Legging Kit with Standard Hood/Voltage Gloves (12)
SDP

Catalog No. 170011491 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1491 3X-Large
17-001-1466 Small
17-001-1471 Medium
17-001-1476 Large
17-001-1481 X-Large
17-001-1486 2X-Large
17-001-1496 4X-Large
17-001-1501 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1491 Supplier National Safety Apparel™ Supplier No. KIT4LC403X12
Add to Cart
Add to Cart

With a unique blend of 88% cotton and 12% high-tenacity nylon, garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability

  • FR hook and loop front closure
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Garments embroidered with arc rating
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Expandable leg opening to fit over boots
  • Hook and loop at side for easy access

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Material Cotton, Nylon
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Gender Unisex
Product Type Arc Flash Kit
Arc Flash Rating 40 cal/cm²
Suspender Adjustable
Size 3X-Large
Closure Type FR Hook and Loop
Color Navy
Seam Type Stitched
Ankle Style Open
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.