Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 65 cal Arc Flash Long Coat/Legging Kit with Standard Hood/Voltage Gloves (10)
SDP

Catalog No. 170011519 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1519 2X-Large
17-001-1504 Small
17-001-1509 Medium
17-001-1514 Large
17-001-1524 3X-Large
17-001-1529 4X-Large
17-001-1536 5X-Large
17-001-1537 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1519 Supplier National Safety Apparel™ Supplier No. KIT4LC652X10
Add to Cart
Add to Cart
Request a Quote

Constructed to be both lightweight and highly durable, this coat and leggings arc flash suit is designed for an arc flash hazard of 65 cal/cm² or less

  • Arc flash resistant caramel brown colored coat and leggings, for complete body protection and preventing heat and burns
  • Arc hood unit with faceshield and hard hat, for head and face protection
  • Hood bag
  • Safety glasses
  • Gear bag
  • 14 in. Class 2 rubber insulating gloves, for hand protection against electric shock up to 17,000 VAC
  • 12 in. leather glove protectors, for protecting the rubber gloves from damage
  • Glove bag for glove storage

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Product Type Arc Flash Kit
Material DuPont™ Nomex™, Kevlar™
Arc Flash Rating 65 cal/cm²
Color Brown
Suspender Adjustable
Closure Type FR Hook and Loop
Ankle Style Open
Size 2X-Large
Seam Type Stitched
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.