Learn More
National Safety Apparel™ ArcGuard™ Performance Arc Flash Kit, 40 Cal
SDP
Catalog No. 17000522 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-522 3X-Large
17-000-518 Small
17-000-519 Large
17-000-520 X-Large
17-000-521 2X-Large
17-000-523 4X-Large
17-000-524 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-000-522 Supplier National Safety Apparel™ Supplier No. ARC40KITNG3X
Add to Cart
Add to Cart
ArcGuard™ Performance Arc Flash Kit, 40 Cal, CAT 4, Includes Jacket, Bib, Lift Front Hood, Backpack
  • Single-layer 40 cal arc flash protection
  • Up to 20% lighter, 48% thinner, and 56% more flexible than other premium arc flash suits
  • Using GORE™ PYRAD™ comfort science to keep the wearer cool, dry, comfortable, and unrestricted while working
  • Ergonomic design for ease of movement and flexibility
  • 26 in. leg zippers for easy donning and doffing
  • Enhanced visibility with 3M™ Scotchlite™ FR silver segmented reflective trim
  • PureView™ face shield technology features a greater range of visibility
  • Gloves are not included
  • Compliant to NFPA 70E for Arc Flash
  • Compliant to CSA Z462 for Arc Flash (Canada)
  • Compliant with OSHA 1910.269
Specifications
Gender Unisex
Suspender Adjustable
Closure Type Zipper, Snap
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, OSHA 1910.269
Color Navy
Material ePTFE (35%), Polyamide (25%), FR Viscose (20%), Aramid (19%), Carbon (1%)
Seam Type Stitched
Size 3X-Large