Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products GL501C5 Series Crinkle Latex-Coated HPPE Gloves

Catalog No. 19555058 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Brown
Red
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
19-555-058 Medium Yellow
19-555-059 Small Red
19-555-057 Large Brown
19-555-061 X-Large Black
19-555-060 2X-Large Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-555-058 Supplier Pyramex Safety Products Supplier No. GL501C5M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Crinkle latex coating on the palm remains flexible over a wide variety of temperatures for enhanced grip capabilities in wet or dry conditions.

  • Crinkle latex palm enhances grip in wet or dry conditions
  • Coating remains flexible and durable over a wide range of temperatures
  • 13-gauge HPPE liner

Specifications

ANSI Abrasion Rating ANSI 4
Coating Material Crinkled Latex
Color Gray, Blue
Cut Resistance Rating ANSI A4
Hand Specific
Material HPPE
Product Type Cut-resistant Gloves
Size Medium
EN388 Standard EN 388:3X42D
For Use With (Application) Glass/material handling, general maintenance, sanitation, wood working
Glove Cuff Color Yellow
Glove Cuff Style Elasticated
Model GL501C5M
Puncture Rating ANSI/ISEA 105-16 Level 4
Series Latex Dipped Series GL501C5
Yarn Gauge 13
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.