Learn More
Radians Arctic Radwear™ Cooling Wrap

Description
- For hot outdoor activities such as construction work, tough workouts or strenuous outdoor/indoor activity
- Soak for 2 to 3 min and wear for up to 5 hrs. of cooling relief (depending on humidity)
- Secure around neck by pulling through slot opening
- Reusable packaging design
- Safe and reusable
- Jobsite proven
- Anti-microbial treated to help prevent mold build-up and unwanted smells
- Heat stress safety management
Specifications
Specifications
| Product Type | Cooling Wrap |
| Color | Hi-Vis Green |
| Dimensions (L x W) | 26 x 8.25 in. |
| For Use With (Application) | For hot outdoor activities such as construction work, tough workouts or any strenuous outdoor or indoor activity |
| Quantity | 10 Pk., 50/Cs. |
Warning: Cancer and reproductive harm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.