Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Vernier Arduino™Interface Shield
SDPSCI_ED

Catalog No. S16105ND Shop All Vernier Software Products
Change view
Click to view available options
Grade:
High School and College
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16105ND High School and College
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16105ND Supplier Vernier™ Supplier No. BTARD
Add to Cart
Add to Cart

Includes

Vernier Arduino Interface Shield, Sample code available online, Arduino is sold separately., Ordering Information, This product is discontinued and no longer available.

The Vernier Arduino Interface Shield provides a convenient way to make connections from Arduino microcontrollers to Vernier sensors.

Vernier Arduino Interface Shield 2 analog sensor ports (BTA sockets) 2 digital sensor ports (BTD sockets) Reset button General-purpose button Indicator LED Includes a multiplexer to allow auto-ID to work on both channels (both analog and digital) Supported Microcontrollers SparkFun Arduino RedBoard Arduino UNO Arduino Leonardo Intel Galileo

Warranty and Services

1 year limited warranty

Specifications

Type Interface
Grade High School and College
Warranty 5 Year Limited
For Use With (Application) Educational use only
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.