Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arecoline Hydrobromide 98.0+%, TCI America™
SDP

Catalog No. A052325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A052325G 25 g
A05235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A052325G Supplier TCI America Supplier No. A052325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 300-08-3
Molecular Formula C8H14BrNO2
Molecular Weight (g/mol) 236.11
MDL Number MFCD00039041
InChI Key AXOJRQLKMVSHHZ-UHFFFAOYSA-N
Synonym arecoline hydrobromide, arecoline bromide, taeniolin, arecoline hbr, methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate hydrobromide, arekolinhydrobromid german, unii-24s79b9cx7, methyl 1,2,5,6-tetrahydro-1-methylnicotinate hydrobromide, n-methyltetrahydronicotinic acid methyl ester hydrobromide
PubChem CID 9301
IUPAC Name hydrogen methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate bromide
SMILES [H+].[Br-].COC(=O)C1=CCCN(C)C1

Specifications

Melting Point 172°C
Quantity 25 g
UN Number 1544
Formula Weight 236.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Arecoline Hydrobromide
RTECSNumber QT2275000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.