Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARF1/ARF3/ARF5/ARF6 Monoclonal Antibody (1D9)
GREENER_CHOICE

Catalog No. MA3060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA3060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA3060 Supplier Invitrogen™ Supplier No. MA3060
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA3-060 detects ADP-ribosylation factor1 (Arf1), Arf3, Arf5 and Arf6 and about ten-fold less well with Arf4 from human, mouse and rat tissues. MA3-060 has been successfully used in Western blot, immunofluorescence, immunocytochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects a single ~21 kDa protein representing Arf in rat pancreas extract. Immunofluorescence staining of Arf in rat pancreas with MA3-060 results in staining of the cytoplasmic face of the trans-Golgi membranes. MA3-060 cannot be used to neutralize Arf in solution. The MA3-060 antigen is recombinant human Arf1.

ADP-ribosylation factor 1 (ARF1) is a member of the humanARFgene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6ARFproteins and 11ARF-like proteins, constitute a family of the RAS superfamily. TheARFproteins are categorized as class I (ARF1,ARF2 andARF3), class II (ARF4 andARF5) and class III (ARF6), and members of each class share a common gene organization. TheARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARF1/ARF3/ARF5/ARF6
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1D9
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.05% sodium azide
Gene Arf4
Gene Accession No. P18085, P61204, P61205, P61206, P61750, P61751, P62330, P62331, P62332, P84077, P84078, P84079, P84082, P84083, P84084, P84085, Q8BSL7
Gene Alias 5430400P17Rik; AA407803; Ac1-253; ADP ribosylation factor 1; ADP ribosylation factor 3; ADP ribosylation factor 4; ADP ribosylation factor 5; ADP ribosylation factor 6; ADP-ribosylation factor 1; ADP-ribosylation factor 2; ADP-ribosylation factor 3; ADP-ribosylation factor 4; ADP-ribosylation factor 4-like protein; ADP-ribosylation factor 5; ADP-ribosylation factor 6; AI788669; AI854770; ARF; Arf1; Arf2; ARF3; ARF4; Arf5; ARF6; AW496366; I79_003112; Liver regeneration-related protein LRRG202; pan ARF; small GTP binding protein
Gene Symbols ARF1, Arf2, ARF3, Arf4, ARF5, ARF6
Host Species Mouse
Immunogen Recombinant human ARF1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100757864, 100759676, 100766631, 100769922, 100774654, 100856517, 11840, 11841, 11842, 11843, 11844, 11845, 140940, 375, 377, 378, 381, 382, 475206, 486562, 490679, 607073, 64310, 79117, 79119, 79120, 79121
Target Species Canine, Hamster, Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.