Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arfaptin-1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501568367
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8367 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8367 Supplier Bethyl Laboratories, Inc Supplier No. A304676AT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Arfaptin 1 is an ARF binding protein that inhibits PLD activation, vesicular trafficking and secretion. In the present report, we show that arfaptin 1 interacts with ′high speed′ membranes independently of ARF. However, addition of myristoylated ARF3 (myrARF3) increases the association of arfaptin 1 with the membranes, suggesting that arfaptin 1 and ARF form a complex on the Golgi. Utilizing several deletion mutants of arfaptin 1 it is shown that the association of arfaptin 1 with myrARF3 is mediated via two binding sites on arfaptin 1. These two domains are needed for arfaptin 1 inhibition of PLD activation by myrARF3 in vitro.

Specifications

Antigen Arfaptin-1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ARFIP1
Gene Accession No. P53367
Gene Alias HSU52521, ADP-ribosylation factor-interacting protein 1, arfaptin 1, arfaptin-1
Gene Symbols ARFIP1
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27236, 99889
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.