Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARFGEF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545214
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545214 Supplier Thermo Scientific Supplier No. PA545214
Only null left
Add to Cart
Add to Cart

ARFGEF1 Polyclonal Antibody

ARFGEF1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ARFGEF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ARFGEF1
Gene Accession No. Q9Y6D6
Gene Alias ADP ribosylation factor guanine nucleotide exchange factor 1; ADP-ribosylation factor guanine nucleotide-exchange factor 1; ADP-ribosylation factor guanine nucleotide-exchange factor 1 (brefeldin A-inhibited); ADP-ribosylation factor guanine nucleotide-exchange factor 1(brefeldin A-inhibited); Arfgef1; ARFGEP1; BIG1; Brefeldin A-inhibited GEP 1; brefeldin A-inhibited guanine nucleotide-exchange protein 1; D130059B05Rik; D730028O18Rik; DKFZP434L057; guanine nucleotide-exchange protein; hm:zeh1651; im:7138204; LOC553514 protein; LOW QUALITY PROTEIN: brefeldin A-inhibited guanine nucleotide-exchange protein 1; P200; p200 ARF guanine nucleotide exchange factor; p200 ARF-GEP1; RGD1560793; wu:fj64h08; zeh1651
Gene Symbols ARFGEF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ARFGEF1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10565
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.