Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Arg-Gly-Asp-Ser

Catalog No. AAJ64877LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64877LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64877LB0 Supplier Thermo Scientific Chemicals Supplier No. J64877LB0
Add to Cart

 

Supports fibroblast attachment

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method HPLC
Common Name Arginyl-glycyl-aspartyl-serine
Content And Storage -30°C to -10°C
Format Powder
For Use With (Application) Supports fibroblast attachment
Molecular Weight (g/mol) 433.42
Product Type Arg-Gly-Asp-Ser
Quantity 5 mg
Regulatory Status RUO - research use only
Sequence RGDS

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.